Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR12 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30943725UL

 View more versions of this product

Catalog No. NB123776


Add to cart

Description

Description

GPR12 Polyclonal antibody specifically detects GPR12 in Human samples. It is validated for Western Blot
Specifications

Specifications

GPR12
Unconjugated
PBS buffer, 2% sucrose
FLJ18149, FLJ97704, G protein-coupled receptor 12, GPCR12, GPCR21, G-protein coupled receptor 12, MGC138349, MGC138351
The immunogen is a synthetic peptide directed towards the C-terminal region of Human GPR12 (NP_005279). Peptide sequence SICLGLLPVMGWNCLRDESTCSVVRPLTKNNAAILSVSFLFMFALMLQLY
25 μg
GPCR
Primary
2835
Western Blot
Western Blot 1.0 ug/ml
Purified
Rabbit
Affinity purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit