Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR128 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5109783
Description
Immunogen sequence: LGIWRIVIRI QRGKSTSSSS TPTEFCRNGG TWENGRCICT EEWKGLRCTI ANFCENSTYM GFTFARIPVG RYGPSLQTCG KDTPNAGNPM AVRLCSLSLY GEIELQKVT.
GPRs are characterized by the presence of 7 transmembrane domains. GPR128 is an orphan receptor.
Specifications
GPR128 | |
Polyclonal | |
Unconjugated | |
ADGRG7 | |
9130020O16Rik; Adgrg7; adhesion G protein-coupled receptor G7; adhesion G-protein coupled receptor G7; FLJ14454; FLJ16382; FLJ29035; G protein-coupled receptor 128; GPR128; G-protein coupled receptor 128; MGC142011; MGC163260; probable G-protein coupled receptor 128 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
239853, 304014, 84873 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.45 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
Q8BM96, Q96K78 | |
ADGRG7 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 27-135 of human GPR128 (NP_116176.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction