Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR128 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13518R

Description
GPRs are characterized by the presence of 7 transmembrane domains. GPR128 is an orphan receptor.Specifications
GPR128 | |
Polyclonal | |
Unconjugated | |
ADGRG7 | |
9130020O16Rik; Adgrg7; adhesion G protein-coupled receptor G7; Adhesion G-protein coupled receptor G7; FLJ14454; FLJ16382; FLJ29035; G protein-coupled receptor 128; GPR128; G-protein coupled receptor 128; MGC142011; MGC163260; probable G-protein coupled receptor 128 | |
Rabbit | |
Protein A | |
RUO | |
84873 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q96K78 | |
ADGRG7 | |
KLH conjugated synthetic peptide derived from mouse G protein coupled receptor 128. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction