Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR132 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA585980
Description
Rabbit Polyclonal Antibody
GPR132 Polyclonal antibody specifically detects GPR132 in Human, Mouse samples. It is validated for Immunocytochemistry, Immunofluorescence, Western BlotSpecifications
GPR132 | |
Polyclonal | |
Unconjugated | |
GPR132 | |
G2 accumulation protein; G2a; Gpr132; MGC99642; Probable G-protein coupled receptor 132 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29933, 56696 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
TBS with 0.5% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4 | |
Q9UNW8, Q9Z282 | |
GPR132 | |
Synthetic peptide corresponding to C terminal of Human GPR132 | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title