Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR133 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen GPR133
Applications Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094131
View Documents
Novus Biologicals
NBP303815100UL
100 μg Each for $320.71
Add to cart
 
NB094132
View Documents
Novus Biologicals
NBP30381520UL
20 μg Each for $138.37
Add to cart
 
Description

Description

GPR133 Polyclonal antibody specifically detects GPR133 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

GPR133
Unconjugated
Rabbit
GPCR
PBS with 50% glycerol, pH7.3.
283383
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot
Purified
RUO
Human, Mouse, Rat
DKFZp434B1272, FLJ16770, G protein-coupled receptor 133, G-protein coupled receptor PGR25, MGC138514, PGR25MGC138512, probable G-protein coupled receptor 133
Recombinant fusion protein containing a sequence corresponding to amino acids 805-874 of human GPR133 (NP_942122.2).
Polyclonal
Affinity purified
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit