Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR142 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548151

Catalog No. PA548151

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

GPR142 is a member of the rhodopsin family of G protein-coupled receptors .
Specifications

Specifications

GPR142
Polyclonal
PBS with 0.09% sodium azide
Q7Z601
Probable G-protein coupled receptor 142, G-protein coupled receptor PGR2, GPR142, PGR2
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
350383
Flow Cytometry, Western Blot
Unconjugated
GPR142
Liquid
GPR142
KLH conjugated synthetic peptide between 86-114 amino acids from the N-terminal region of human GPR142.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit