Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR160 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP309485100UL
Description
GPR160 Polyclonal specifically detects GPR160 in Human samples. It is validated for Western Blot.Specifications
GPR160 | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
G protein-coupled receptor 160, GPCR150GPCR1, G-protein coupled receptor GPCR1, hGPCR1, probable G-protein coupled receptor 160, putative G protein-coupled receptor | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human GPR160 (NP_055188). Peptide sequence FSSHSSYTVRSKKIFLSKLIVCFLSTWLPFVLLQVIIVLLKVQIPAYIEM | |
100 μg | |
GPCR | |
26996 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
PBS buffer, 2% sucrose | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction