Learn More
MBL International Corporation GPR17 50 UL DFS Item
Manufacturer:  MBL International Corporation MC4229
G Protein-Coupled Receptor GPR17 Polyclonal Antibody Size 50 ul Clone Number Polyclonal Host Species Rabbit Conjugate Application ICC , IHC
Catalog No. 50-489-49