Learn More
MBL International Corporation G Protein-Coupled Receptor AGR9 Polyclonal Antibody (US Only)
Supplier:  MBL International Corporation MC401
Encompass_Preferred
Clonality:Polyclonal; Application:IHC; Conjugate:Unlabeled; Species Reactivity:Human; Size:50 µl; Target:GPR176; GPR176 Primary Antibody of 50 µl. For sale In the US only.GPR176 Primary Antibody.
Catalog No. 50-489-04