Learn More
GPR21 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 179731AP
Catalog No. 50-173-1909
Add to cart
Description
Description
Human follicle stimulating hormone is a glycoprotein produced from urine of post-menopausal women and having a total molecular mass of 35,500 Dalton. HuFSH is a heterodimeric hormone consisting of 165 amino acids a chain (non-active) and 115 amino acids b chain (active). HuFSH induces maturation of Graafian follicles of the ovary, promotes development of germinal cells in males and activates cytosolic tyrosine kinase. The HuFSH is purified by proprietary chromatographic techniques.
Specifications
Specifications
GPR21
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q8BX79, Q99679
G protein coupled receptor 21, GPR21
Rabbit
IgG
150 μL
-20°C
Primary
2844, 338346, 689917
Immunohistochemistry (Paraffin)
Unconjugated
GPR21
Liquid
GPR21
GPR21 Fusion Protein Ag12064
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat