Learn More
GPR26 Mouse anti-Human, Alexa Fluor 647, Clone: 891318, R&D Systems™
Mouse MonoclonalAntibody
Manufacturer:  R&D Systems FAB8449R100UG
Catalog No. FAB8449R100
Add to cart
This item is not returnable. View return policy
Description
Description
GPR26 Monoclonal antibody specifically detects GPR26 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GPR26
Monoclonal
0.2 mg/mL
Flow Cytometry 0.25-1 ug/10^6 cells
Q8NDV2
GPR26
NS0 mouse myeloma cell line transfected with human GPR26 Met1-Glu337 Accession # Q8NDV2
100 ug
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date.
Monoclonal
Stains human GPR26 transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
891318
Alexa Fluor 647
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide
DKFZp761H2121, FLJ33774, G protein-coupled receptor 26, G-protein coupled receptor 26, MGC138216
Mouse
IgG2b
RUO
Primary
2849
Human
SDS
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)