Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR37L1 Antibody (444532), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4449N
Description
GPR37L1 Monoclonal antibody specifically detects GPR37L1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
GPR37L1 | |
Monoclonal | |
Alexa Fluor 700 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
444532 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
endothelin B receptor-like protein 2, ET(B)R-LP-2, ETBRLP2, ETBR-LP-2endothelin type b receptor-like protein 2, G protein-coupled receptor 37 like 1, G-protein coupled receptor 37 like 1, G-protein coupled receptor 37-like 1 | |
NS0 mouse myeloma cell line transfected with human GPR37L1, Accession # O60883 | |
0.1 mL | |
GPCR, Neuroscience, Signal Transduction | |
9283 | |
Store at 4C in the dark. | |
IgG2a |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction