Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ GPR39 (Human) Recombinant Protein

For use with Antibody Production.

Manufacturer:  Abnova™ H00002863G01

Catalog No. 89-339-390


Add to cart

Description

Description

Human GPR39 full-length ORF (NP_001499.1) recombinant protein without tag.
Specifications

Specifications

Antibody Production
G protein-coupled receptor 39
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
2863
10 µg
MGC149541
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit