Learn More
MBL International Corporation G Protein-Coupled Receptor GPR44 (CRTH2) Polyclonal Antibody (US Only)
Supplier:  MBL International Corporation MC3528
Encompass_Preferred
Clonality:Polyclonal; Application:ICC|IHC; Conjugate:Unlabeled; Species Reactivity:Human; Size:50 µl; Target:GPR44; GPR44 Primary Antibody of 50 µl. For sale In the US only.GPR44 Primary Antibody.
Catalog No. 50-488-26