Learn More
GPR64 Mouse anti-Human, Alexa Fluor 405, Clone: 206420, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB7977V100UG
Catalog No. FAB7977V100
Add to cart
This item is not returnable. View return policy
Description
Description
GPR64 Monoclonal antibody specifically detects GPR64 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GPR64
206420
Flow Cytometry 0.25-1 μg/106 cells
NP_001073328
EDDM6, epididymis-specific (seven transmembrane family), G protein-coupled receptor 64, HE6, MGC104454, MGC138738, TM7LN2
Chinese hamster ovary cell line CHO-derived recombinant human GPR64, Leu38-Asn64, Glu68-Thr553, Accession No. NP_001073328
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human GPR64 transfectants in cell-based ELISA and Flow Cytometry.
Flow Cytometry
Alexa Fluor 405
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
10149.0
Human