Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR74/NPFFR2 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$204.00 - $409.50

Specifications

Antigen GPR74/NPFFR2
Concentration 1 mg/mL
Dilution Western Blot 1:500-1:1000
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB019389
SDP
View Documents
Novus Biologicals
NBP224879SS
0.025 mg
Each for $204.00
Add to Cart
 
NBP224879
SDP
View Documents
Novus Biologicals
NBP224879
0.1 mg
Each for $409.50
Add to Cart
 
Description

Description

GPR74/NPFFR2 Polyclonal specifically detects GPR74/NPFFR2 in Human samples. It is validated for Western Blot.
Specifications

Specifications

GPR74/NPFFR2
Western Blot 1:500-1:1000
Polyclonal
Rabbit
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission
PBS with 0.02% Sodium Azide
10886
Reacts with the human NPFF2 protein.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
1 mg/mL
Western Blot
Unconjugated
RUO
Human
GPR74G protein-coupled receptor 74, G-protein coupled receptor 74, neuropeptide FF 2, neuropeptide FF receptor 2, Neuropeptide G-protein coupled receptor, NPFF2G-protein coupled receptor HLWAR77, NPGPRHLWAR77
NPFFR2
IgG
Affinity Purified
SDS
Documents

Documents

Product Certifications

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit