Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR74/NPFFR2 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
$204.00 - $409.50
Specifications
Antigen | GPR74/NPFFR2 |
---|---|
Concentration | 1 mg/mL |
Dilution | Western Blot 1:500-1:1000 |
Applications | Western Blot |
Classification | Polyclonal |
Description
GPR74/NPFFR2 Polyclonal specifically detects GPR74/NPFFR2 in Human samples. It is validated for Western Blot.Specifications
GPR74/NPFFR2 | |
Western Blot 1:500-1:1000 | |
Polyclonal | |
Rabbit | |
GPCR, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
PBS with 0.02% Sodium Azide | |
10886 | |
Reacts with the human NPFF2 protein. | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
1 mg/mL | |
Western Blot | |
Unconjugated | |
RUO | |
Human | |
GPR74G protein-coupled receptor 74, G-protein coupled receptor 74, neuropeptide FF 2, neuropeptide FF receptor 2, Neuropeptide G-protein coupled receptor, NPFF2G-protein coupled receptor HLWAR77, NPGPRHLWAR77 | |
NPFFR2 | |
IgG | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title