Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPR77 293T Cell Overexpression Lysate (Denatured), Abnova

Cell transient overexpression lysate for GPR77

Manufacturer:  Abnova Corporation H00027202T01

Catalog No. 89-002-109


Add to cart

Description

Description

The anaphylatoxins C3a, C4a, and C5a are cationic fragments generated during the complement cascade that participate in host defense. In the case of inappropriate complement activation, anaphylatoxins may be involved in autoimmunity and sepsis. C5L2 is coexpressed with the C5a receptor, C5AR (C5R1), on polymorphonuclear neutrophils and may modulate C5AR activity.

  • G protein-coupled receptor 77
  • Transfected Cell Line: 293T
  • Plasmid: pCMV-GPR77 full-length
  • Host: Human
  • Theoretical MW: 36.1kDa
  • Storage Buffer: 1X Sample Buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% Bromophenol blue)

Western Blotting

Specifications

Specifications

Western Blot
293T
NM_018485.1
6.8
Transient overexpression cell Lysate was tested with Anti-GPR77 antibody (H00027202-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
GPR77 293T Cell Transient Overexpression Lysate(Denatured)
36.1
pCMV-GPR77 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit