Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR83 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA533770
Description
Percent identity with other species by BLAST analysis: Human (93%) Chimpanzee, Gorilla, Orangutan, Monkey, Mouse (87%) Gibbon, Galago, Marmoset, Rat, Bat, Hamster, Rabbit, Horse (80%).
GPR83, a 423 A.A protein, is an orphan member of GPCR protein family and is closely related to tachykinin and NPY receptors. It is a human homolog of mouse GIR protein and is exclusively expressed and selectively up-regulated in brain. It is a cell surface receptor protein consisting of a putative seven-transmembrane-domain. Reports suggest GPR83 might be involved in the regulation of immune tolerance mechanism by inducing peripheral generation of Foxp3+ Treg cells.
Specifications
GPR83 | |
Polyclonal | |
Unconjugated | |
GPR83 | |
AW045253; G protein-coupled receptor 72; G protein-coupled receptor 83; Gir; glucocorticoid induced receptor; glucocorticoid-induced receptor; GPR72; Gpr83; G-protein coupled receptor 72; Hcepto9; Jp05; KIAA1540; Probable G-protein coupled receptor 83; RP105; RP39; RP82 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10888 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9NYM4 | |
GPR83 | |
Synthetic 16 amino acid peptide from C-terminus of human GPR83. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction