Learn More
GPR83 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 238501AP
Catalog No. 50-173-4142
Add to cart
Description
Description
GPR83, a 423 A.A protein, is an orphan member of GPCR protein family and is closely related to tachykinin and NPY receptors. It is a human homolog of mouse GIR protein and is exclusively expressed and selectively up-regulated in brain. It is a cell surface receptor protein consisting of a putative seven-transmembrane-domain. Reports suggest GPR83 might be involved in the regulation of immune tolerance mechanism by inducing peripheral generation of Foxp3+ Treg cells.
Specifications
Specifications
GPR83
0.55 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NYM4
G protein coupled receptor 72, G protein coupled receptor 83, GIR, GPR72, GPR83, KIAA1540
Rabbit
IgG
150 μL
-20°C
Primary
10888
Western Blot
Unconjugated
GPR83
Liquid
GPR83
GPR83 Fusion Protein Ag20873
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human