Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GPR87 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156442
Description
GPR87 Polyclonal specifically detects GPR87 in Human samples. It is validated for Western Blot.Specifications
GPR87 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
G protein-coupled receptor 87, G protein-coupled receptor 95, GPR95, G-protein coupled receptor 87, G-protein coupled receptor 95, KPG_002, MGC131898, orphan GPCR 87 | |
Rabbit | |
41 kDa | |
100 μL | |
GPCR | |
53836 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml | |
Q9BY21 | |
GPR87 | |
Synthetic peptides corresponding to GPR87 (G protein-coupled receptor 87) The peptide sequence was selected from the middle region of GPR87)(50ug). Peptide sequence NQSIRVVVAVFFTCFLPYHLCRIPFTFSHLDRLLDESAQKILYYCKEITL. | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Chicken: 78%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction