Learn More
GPR97 Rabbit anti-Human, Alexa Fluor™ 700, Clone: 2356C, R&D Systems™
Rabbit Monoclonal Antibody
Manufacturer:  R&D Systems FAB10097N100UG
Catalog No. FB10097N100
Add to cart
This item is not returnable. View return policy
Description
Description
GPR97 Monoclonal antibody specifically detects GPR97 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GPR97
2356C
Flow Cytometry 0.25-1 μg/106 cells
Q86Y34
Rabbit
IgG
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human GPR97 in direct ELISAs.
Flow Cytometry
Alexa Fluor™ 700
Supplied 0.2 mg/mL in a saline solution containing BSA and sodium azide
G protein-coupled receptor 97, Pb99, PGR26, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97
Human embryonic kidney cell, HEK293-transfected human GPR97, Met1-Leu249, Accession No. Q86Y34
100 μg
Primary
222487
Human