Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPR98 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA599894
Description
Antibody detects endogenous levels of total GPR98.
This gene encodes a member of the G-protein coupled receptor superfamily. The encoded protein contains a 7-transmembrane receptor domain, binds calcium and is expressed in the central nervous system. Mutations in this gene are associated with Usher syndrome 2 and familial febrile seizures. Several alternatively spliced transcripts have been described.
Specifications
GPR98 | |
Polyclonal | |
Unconjugated | |
ADGRV1 | |
4-Feb; ADGRV1; ADGRV1 subunit alpha; ADGRV1 subunit beta; adhesion G protein-coupled receptor V1; Adhesion G-protein coupled receptor V1; FEB4; Frings; G protein-coupled receptor 98; Gpr98; G-protein coupled receptor 98; KIAA0686; KIAA1943; MASS1; Mgr1; monogenic; monogenic audiogenic seizure susceptibility protein 1; monogenic audiogenic seizure susceptibility protein 1 homolog; monogenic, audiogenic seizure susceptibility 1; monogenic, audiogenic seizure susceptibility 1 homolog; neurepin; USH2B; USH2C; Usher syndrome type-2C protein; very large G protein-coupled receptor 1; very large G-protein coupled receptor 1; VLGR1; VLGR1b | |
Rabbit | |
Affinity Chromatography | |
RUO | |
110789, 84059 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q8WXG9 | |
ADGRV1 | |
A synthesized peptide derived from human GPR98(Accession Q8WXG9), corresponding to amino acid residues L2004-S2054. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction