Learn More
GPRASP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 200201AP
Catalog No. 50-173-2657
Add to cart
Description
Description
The antibody is specific to GPRASP1.
Modulates lysosomal sorting and functional down-regulation of a variety of G-protein coupled receptors. Targets receptors for degradation in lysosomes via its interaction with BECN2.
Specifications
Specifications
GPRASP1
0.19 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q5JY77, Q5U4C1
GASP, GASP 1, GASP1, GPRASP1, KIAA0443
Rabbit
IgG
150 μL
-20°C
Primary
67298, 9737
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
GPRASP1
Liquid
Gprasp1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse