Learn More
GPRASP2 Mouse anti-Human, Alexa Fluor™ 350, Clone: MM0304-6M51, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212327AF350
Catalog No. NP212327A30
Add to cart
Description
Description
GPRASP2 Monoclonal antibody specifically detects GPRASP2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
GPRASP2
MM0304-6M51
Western Blot
FLJ35662, FLJ37327, G protein-coupled receptor associated sorting protein 2, GASP2, GASP-2, G-protein coupled receptor-associated sorting protein 2
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
GPRASP2
Human GPRASP2
0.1 mL
Primary
114928