Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC5D 293T Cell Overexpression Lysate (Denatured), Abnova

Cell transient overexpression lysate for GPRC5D

Manufacturer:  Abnova Corporation H00055507T01

Catalog No. 89-002-439


Add to cart

Description

Description

The protein encoded by this gene is a member of the G protein-coupled receptor family; however, the specific function of this gene has not yet been determined.

  • G protein-coupled receptor, family C, group 5, member D
  • Transfected cell line: 293T
  • Plasmid: pCMV-GPRC5D full-length
  • Host: Human
  • Theoretical molecular weight: 38.06kDa
  • Storage buffer: 1X sample buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)

Western blotting

Specifications

Specifications

Western Blot
293T
NM_018654
6.8
Transient overexpression cell Lysate was tested with Anti-GPRC5D antibody (H00055507-B01) by western blotting
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human
GPRC5D 293T Cell Transient Overexpression Lysate(Denatured)
38.8
pCMV-GPRC5D full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit