Learn More
GPRC5D Mouse anti-Human, Alexa Fluor 700, Clone: 571961, R&D Systems
Mouse Monoclonal Antibody
Manufacturer:  R&D Systems FAB6300N100UG
Catalog No. FB6300N100
Add to cart
This item is not returnable. View return policy
Description
Description
GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
GPRC5D
571961
Flow Cytometry 0.25-1 μg/106 cells
Q9NZD1
G protein-coupled receptor, family C, group 5, member D, G-protein coupled receptor family C group 5 member D, MGC129713, MGC129714, orphan G-protein coupled receptor
NS0 mouse myeloma cell line transfected with human GPRC5D, Met1-Val359, Accession No. Q9NZD1
Protein A or G purified from hybridoma culture supernatant
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied.
Monoclonal
Detects human GPRC5D in direct ELISAs. Stains human GPRC5D transfectants but not irrelevant transfectants in flow cytometry.
Flow Cytometry
Alexa Fluor 700
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Purified, Supernatant, Conjugated
Mouse
IgG2b
100 μg
Primary
55507.0
Human