Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GPRC5D Mouse anti-Human, Clone: 571961, R&D Systems™ Available on GSA/VA Contract for Federal Government customers only.

Mouse MonoclonalAntibody

$116.82 - $301.00

Specifications

Antigen GPRC5D
Clone 571961
Dilution Flow Cytometry 0.25 ug/10^6 cells
Applications Flow Cytometry
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Conjugate Price Quantity & Availability  
MAB6300100 Available on GSA/VA Contract for Federal Government customers only. View Documents R&D Systems
MAB6300100
100 ug Unconjugated Each for $301.00
Add to cart
 
MAB6300SP View Documents R&D Systems
MAB6300SP
25 ug Unconjugated Each for $116.82
Add to cart
 
Description

Description

GPRC5D Monoclonal antibody specifically detects GPRC5D in Human samples. It is validated for Flow Cytometry.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

GPRC5D
Flow Cytometry 0.25 ug/10^6 cells
Purified
Human
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
NS0 mouse myeloma cell line transfected with human GPRC5D Met1-Val359 Accession # Q9NZD1
Monoclonal
Protein A or G purified from hybridoma culture supernatant
Detects human GPRC5D in direct ELISAs. Stains human GPRC5D transfectants but not irrelevant transfectants in flow cytometry.
571961
Flow Cytometry
Mouse
RUO
55507
IgG2b
Primary
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.

For Research Use Only (RUO)

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit