Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ GPRIN1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550034
Description
The antibody detects endogenous levels of total GPRIN1 protein.
May be involved in neurite outgrowth.
Specifications
GPRIN1 | |
Polyclonal | |
Unconjugated | |
GPRIN1 | |
G protein regulated inducer of neurite outgrowth 1; G protein-regulated inducer of neurite outgrowth 1; Gprin1; GRIN1; KIAA1893; Z16 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
114787 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q7Z2K8 | |
GPRIN1 | |
Synthesized peptide derived from internal of human GPRIN1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction