Learn More
Aviva Systems Biology Corp GPT2 Antibody - C-terminal region (ARP48996_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP48996_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GPT2. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human GPT2. Alternate names: ALT2ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59988