Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ly-6G/Ly-6C Rat anti-Mouse, PE, Clone: RB6-8C5, Invitrogen™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen RM3004

Catalog No. 50-113-7585


Add to cart

Description

Description

Granulocytes are a category of white blood cells characterised by the presence of granules in their cytoplasm. They are also called polymorphonuclear leukocytes (PMN or PML) because of the varying shapes of the nucleus, which is usually lobed into three segments. In common parlance, the term polymorphonuclear leukocyte often refers specifically to neutrophil granulocytes, the most abundant of the granulocytes. Granulocytes or PMN are released from the bone marrow by the regulatory complement proteins.
Specifications

Specifications

Ly-6G/Ly-6C, GR-1, LY-6C/G
Monoclonal
0.2 mg/mL
PBS with 4mg/mL BSA, sucrose and 0.1% sodium azide
P35461
Gr-1, Gr1, Ly-6G, ly-6G.1, lymphocyte antigen 6G
Rat
IgG2b
500 μL
4° C
Primary
546644
Flow Cytometry
RB6-8C5
PE
Ly6g
Liquid
Ly6g
Normal murine bone marrow cells
Purified
RUO
Antibody
Monoclonal
Mouse
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit