Learn More
Gr-1/Ly-6G Rat anti-Mouse, Alexa Fluor™ 750, Clone: 1A8, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP253131AF750
Catalog No. NP23131A70
Add to cart
Description
Description
Gr-1/Ly-6G Monoclonal antibody specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Gr-1/Ly-6G
1A8
Flow Cytometry
C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25
Rat
IgG2a Kappa
Store at 4°C in the dark.
Monoclonal
Mouse
Flow Cytometry
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
LY6G6D
Ly-6G transfected EL-4J cell line.
0.1 mL
Primary
58530