Learn More
GR 103691, Tocris
Highly selective D3 antagonist
Manufacturer:  R&D Systems 1109/10
Quantity 10mg
 View more versions of this product
Catalog No. 11-091-0
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
>98%
485.63
GR 103691
C30H35N3O3
4'-Acetyl-N-[4-[4-(2-methoxyphenyl)-1-piperazinyl]butyl]-[1, 1'-biphenyl]-4-carboxamide
Specifications
162408-66-4
C30H35N3O3
4'-Acetyl-N-[4-[4-(2-methoxyphenyl)-1-piperazinyl]butyl]-[1, 1'-biphenyl]-4-carboxamide
GR 103691
>98%
485.63
10mg
Safety and Handling
Recommended Storage : Store at Room Temperature