Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ GR 125487 sulfamate
GREENER_CHOICE

Catalog No. p-4783489
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective 5-HT4 antagonist; active in vivo

Potent and selective 5-HT4 receptor antagonist (Ki = 0.19 nM for porcine 5-HT4). Centrally active following systemic administration in vivo.

Chemical Identifiers

CAS 859502-43-5
Molecular Formula C19H29FN4O8S2

Specifications

Quantity 10 mg
Formula Weight 524.58
Purity >98%
Molecular Weight (g/mol) 524.579
Chemical Name or Material GR 125487 sulfamate
Recommended Storage Desiccate at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.