Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GR 135531, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Binds with high affinity to MT3 melatonin sites in hamster brain, and low affinity (and no efficacy) at MT1 and MT2 melatonin receptors and thus can be used to discriminate between the melatonin receptor subtypes.
Specifications
Quantity | 10 mg |
Formula Weight | 275.31 |
Percent Purity | >97% |
Chemical Name or Material | GR 135531 |
Safety and Handling
Recommended Storage | Store at Room Temperature |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction