GR 144053 trihydrochloride, Tocris

Glycoprotein IIb/IIIa (integrin αIibβ3) receptor antagonist; Antithrombotic

Manufacturer: R&D Systems 1263/50

Quantity 50mg

 View more versions of this product

Catalog No. 12-635-0

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >99%
    Molecular Formula C18H27N5O2·3HCl
    Formula Weight 454.83
    Synonym 4-[4-[4-(Aminoiminomethyl)phenyl]-1-piperazinyl]-1-piperidineacetic acid trihydrochloride
    Chemical Name or Material GR 144053 trihydrochloride

    Specifications

    Formula Weight 454.83
    Quantity 50mg
    Assay Percent Range >99%
    Molecular Formula C18H27N5O2·3HCl
    CAS 1215333-48-4
    Synonym 4-[4-[4-(Aminoiminomethyl)phenyl]-1-piperazinyl]-1-piperidineacetic acid trihydrochloride
    Recommended Storage Desiccate at +4°C
    Chemical Name or Material GR 144053 trihydrochloride

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit