Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GR 159897, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
A potent, selective, non-peptide, orally active neurokinin NK2 receptor antagonist. Competes for binding of [3H]GR100679 to hNK2-transfected CHO cells with a pKi of 9.5. Inhibits NK2 receptor-mediated contraction of guinea pig trachea with a pA2 of 8.7. Anxiolytic in vivo.
Specifications
Quantity | 50 mg |
Formula Weight | 414.54 |
Percent Purity | >97% |
Chemical Name or Material | GR 159897 |
Safety and Handling
Recommended Storage | Desiccate at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction