Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GR 159897, Tocris Bioscience™

Catalog Number p-4783494
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
158848-32-9
C23H27FN2O2S
414.539
BANYJBHWTOJQDU-GDLZYMKVSA-N
5-fluoro-3-2-4-methoxy-4-r-phenylsulphinyl methyl-1-piperidinyl ethyl-1h-indole, d02ywv, r-5-fluoro-3-2-4-methoxy-4-phenylsulfinyl methyl piperidin-1-yl ethyl-1h-indole, 5-fluoro-3-2-4-methoxy-4-r-phenylsulfinyl methyl piperidin-1-yl ethyl-1h-indole, 5-fluoro-3-2-4-methoxy-4-r phenylsulfinyl methyl-1-piperidinyl ethyl-1h-indole, r-1-2-5-fluoro-1h-indol-3-yl ethyl-4-methoxy-4-phenyl-sulfinyl methyl-piperidine, 3-2-4-r-benzenesulfinyl methyl-4-methoxypiperidin-1-yl ethyl-5-fluoro-1h-indole
9953599
5-fluoro-3-[2-[4-methoxy-4-[[(R)-phenylsulfinyl]methyl]piperidin-1-yl]ethyl]-1H-indole
COC1(CCN(CC1)CCC2=CNC3=C2C=C(C=C3)F)CS(=O)C4=CC=CC=C4
This item is not returnable. View return policy
158848-32-9
C23H27FN2O2S
414.539
BANYJBHWTOJQDU-GDLZYMKVSA-N
5-fluoro-3-2-4-methoxy-4-r-phenylsulphinyl methyl-1-piperidinyl ethyl-1h-indole, d02ywv, r-5-fluoro-3-2-4-methoxy-4-phenylsulfinyl methyl piperidin-1-yl ethyl-1h-indole, 5-fluoro-3-2-4-methoxy-4-r-phenylsulfinyl methyl piperidin-1-yl ethyl-1h-indole, 5-fluoro-3-2-4-methoxy-4-r phenylsulfinyl methyl-1-piperidinyl ethyl-1h-indole, r-1-2-5-fluoro-1h-indol-3-yl ethyl-4-methoxy-4-phenyl-sulfinyl methyl-piperidine, 3-2-4-r-benzenesulfinyl methyl-4-methoxypiperidin-1-yl ethyl-5-fluoro-1h-indole
9953599
5-fluoro-3-[2-[4-methoxy-4-[[(R)-phenylsulfinyl]methyl]piperidin-1-yl]ethyl]-1H-indole
COC1(CCN(CC1)CCC2=CNC3=C2C=C(C=C3)F)CS(=O)C4=CC=CC=C4

Non-peptide, potent NK2 antagonist

A potent, selective, non-peptide, orally active neurokinin NK2 receptor antagonist. Competes for binding of [3H]GR100679 to hNK2-transfected CHO cells with a pKi of 9.5. Inhibits NK2 receptor-mediated contraction of guinea pig trachea with a pA2 of 8.7. Anxiolytic in vivo.
Quantity 10 mg
Formula Weight 414.54
Percent Purity >97%
Chemical Name or Material GR 159897
Recommended Storage Desiccate at -20°C
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit