Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GR 79236, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
124555-18-6
C15H21N5O5
351.363
GYWXTRVEUURNEW-KOALMESCSA-N
N-[(1S, 2S)-2-Hydroxycyclopentyl]adenosine
16759592
(2S,3R,4S,5R)-2-[6-[[(1S,2S)-2-hydroxycyclopentyl]amino]purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
C1CC(C(C1)O)NC2=NC=NC3=C2N=CN3C4C(C(C(O4)CO)O)O
This item is not returnable. View return policy
124555-18-6
C15H21N5O5
351.363
GYWXTRVEUURNEW-KOALMESCSA-N
N-[(1S, 2S)-2-Hydroxycyclopentyl]adenosine
16759592
(2S,3R,4S,5R)-2-[6-[[(1S,2S)-2-hydroxycyclopentyl]amino]purin-9-yl]-5-(hydroxymethyl)oxolane-3,4-diol
C1CC(C(C1)O)NC2=NC=NC3=C2N=CN3C4C(C(C(O4)CO)O)O

A1 agonist

Adenosine A1 receptor agonist (Ki = 3.1 nM). Anticonvulsive in mice following systemic administration in vivo.
Quantity 50mg
Formula Weight 351.36
Percent Purity >99%
Chemical Name or Material GR 79236

Recommended Storage : Desiccate at +4°C