Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GR/NR3C1 Rabbit anti-Human, Mouse, Rat, Fish, Zebrafish, Clone: JF0952, Novus Biologicals™
Rabbit Monoclonal Antibody
Manufacturer: Novus Biologicals NBP266878
Description
GR/NR3C1 Monoclonal antibody specifically detects GR/NR3C1 in Human, Mouse, Rat, Fish, Zebrafish samples. It is validated for Western Blot, Flow Cytometry.Specifications
GR/NR3C1 | |
JF0952 | |
Unconjugated | |
Purified | |
GCR, glucocorticoid receptor, GRGCCR, GRLNuclear receptor subfamily 3 group C member 1, nuclear receptor subfamily 3, group C, member 1, nuclear receptor subfamily 3, group C, member 1 (glucocorticoid receptor) | |
NR3C1 | |
Recombinant Protein | |
Monoclonal | |
Protein A purified | |
Phospho Specific, Signal Transduction | |
Human,Mouse,Rat,Fish,Zebrafish |
Flow Cytometry,Western Blot | |
1 mg/mL | |
Western Blot 1:1000, Flow Cytometry 1:50-1:100 | |
TBS (pH7.4), 1%BSA, 40%Glycerol with 0.05% Sodium Azide | |
2908 | |
Rabbit | |
IgG | |
Primary | |
100μL | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title