Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grace™ Vydac™ 214 MS C4 Analytical HPLC Columns, 5µm Particle Size

Suitable for small peptides to large intact, undenatured proteins

Manufacturer:  Grace™ 5103899

 View more versions of this product

Catalog No. 07-650-053

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications Description & Specifications

Specifications

3%
Yes
1mL/min.
250mm
0.1% v/v TFA in Water; 0.085% v/v TFA in Acetonitrile
Spherical
300Å
25°C
L26
Vydac™ 214 MS C4
4.6mm
5μm
LC/MS separation
1000 to 2000psi
Silica
5μm
70 to 110m2/g
HPLC Column
Reverse phase HPLC
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit