Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Grace™ Vydac™ 214 MS C4 Analytical HPLC Columns, 5µm Particle Size

Suitable for small peptides to large intact, undenatured proteins

Manufacturer:  Grace™ 5103897

 View more versions of this product

Catalog No. 07-650-049

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Specifications Description & Specifications

Specifications

3%
2.1mm
5μm
50mm
Spherical
Polymeric
70 to 110m2/g
L26
Vydac™ 214 MS C4
Vydac™ 214 MS C4
Yes
LC/MS separation
Silica
5μm
300Å
HPLC Column
Reverse phase HPLC
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit