Learn More
Grace™ Vydac™ 218 MS Polymeric C18 Analytical HPLC Columns
Suitable for small peptides to large intact, undenatured proteins
Manufacturer:  Grace™ 5103966
 View more versions of this product
Catalog No. 07-650-112
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
8%
4.6mm
5μm
150mm
Spherical
Polymeric
60 to 110m2/g
HPLC Column
Reverse phase HPLC
Vydac™ 218 MS C18
Yes
LC/MS separation
Silica
5μm
300Å
22°C
L1
Vydac™ 218 MS C18