Learn More
AIMS™ Grace Procedure/Dissection Board Products for Science Education
For small animal surgery, procedure, and dissection platforms.
Manufacturer:  AIMS™ GPM2
 View more versions of this product
Catalog No. 14-370-214
Add to cart
Description
Description
  • Construction: High Density Polyethylene (HDPE) with excellent chemical resistance
  • Corner magnets for holding metal accessories
  • Recessed Steel Bars for use with magnetic accessories
  • Flexible LED lights - provides direct illu
Specifications
Specifications
Use for dissection of small animals
Two LED lights, magnifier and magnetic base system
HDPE, stainless steel (warming plate)
100 to 240V 50 to 60Hz
Surgical board
2.3 X (main lens); 4.0X (inset lens)
30.48 x 24.82cm
Mouse, rats