Learn More
BioworldSupplier Diversity Partner Grace's Suppl Media (Hink's), 500 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306110331
Grace's Insect Cell Culture Media is a modification of Wyatt's medium and was originally formulated to support the growth of cells derived from the Australian emperor gum moth, Anthere eucalypti. When supplemented with the proper components, this medium has been shown to support a variety of insect cells, including several lepidopteran as well as some dipterans. Although it has been shown to support the growth of some dipterans, it is primarily used in the cultivation of cell lines derived from lepidopterans. Grace's Supplement Media (Hink's TNM-FH) item # 785075, contains L-glutamin, 3330 mg/L lactabumin hydrosulfate and 3330 mg/L yeastolate. Click here to see a Grace's Insect Medium Components Table.
Catalog No. 50-254-085