Grace™ Vydac™ C18 (218TP) HPLC Columns

Designed for the separation of peptides less than 4000 to 5000 MW and enzymatic digest fragments

Manufacturer: Grace™ 5103984

 View more versions of this product

Catalog No. 07-650-116

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Particle Size 5μm
    Pore Size 300Å
    For Use With LC/MS separation
    Stationary Phase Vydac™ 218TP C18
    Type HPLC Column
    Length (Metric) 250mm
    Support Material Silica
    Diameter (Metric) Inner 10mm
    Phase Polymeric
    Column Type Vydac™ 218TP C18
    Flow Rate 5mL/min.
    Packing Material Vydac™ 218TP C18
    Technique Reverse phase HPLC
    Carbon Load 8%
    USP Type L1
    Endcapped Yes

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit