Learn More
BUCHI FlashPure Flash Chromatography Cartridges
FlashPure flash chromatography cartridges offer high resolution and outstanding loading capacity.
Manufacturer:  BUCHI 11067708
 View more versions of this product
Catalog No. 05-404-815
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Includes: 10 Pieces
Description
Description
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
Specifications
Specifications
Silica - Normal Phase
10 Pc.
For Flash Chromatography Using ISCO and Biotage Instruments
FlashPure
120 g
75 psig