Learn More
BUCHI FlashPure Silica Flash Cartridges Available on GSA/VA Contract for Federal Government customers only.
High resolution and outstanding loading capacity
Manufacturer:  BUCHI 11067710
 View more versions of this product
Catalog No. 05-404-816
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: 4 Pieces
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
330 g
4 Pc.
75 psig
For Flash Chromatography Using ISCO and Biotage Instruments
Silica - Normal Phase
FlashPure