Learn More
BUCHI GraceResolv™ LOK Flash Cartridges
Supplied with an adapter that allows connection to systems using a luer slip outlet for maximum flexibility
Manufacturer:  BUCHI 145171340
 View more versions of this product
Catalog No. 05-404-849
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: 20 Pieces
Description
Description
High performance
  • High resolution of separation due to high-quality silica
  • Increased fraction purity due to a better separation
  • Faster evaporation of smaller fraction volumes
  • Reproducible performance from batch to batch
Lower purification costs
  • Increased sample throughput due to high sample loading capacity
  • Solvent saving: required purification yield is achieved faster
  • /ul>
Specifications
Specifications
4 g
20 Pc.
75 psig
Supplied with an Adapter That Allows Connection to Systems Using a Luer Slip Outlet
Silica - GraceResolve
GraceResolv