Learn More
BioworldSupplier Diversity Partner Grace's Insect Medium, 500 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306110961
Grace’s insect medium is ideal for transfection when supplemented with 2-10% fetal bovine serum.It supports a variety of insect cells, including several lepidopterans and some dipterans.This medium lacks the lactalbumin hydrolysate and yeastolate added to TNM-FH medium. We manufacture all our insect media only with stabilized L-glutamine to ensure the longest possible shelf life.SpecificationsSterile-filteredShips on ice.
Catalog No. 50-254-213